Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006699.1 | NP_001006700.1 | 9 - 10 | 1046 | 92 | |
XM_012953020.2 | XP_012808474.1 | 10 - 11 | 1248 | 92 | ||
XM_012953021.2 | XP_012808475.1 | 8 - 9 | 877 | 92 | ||
GenBank mRNA | BC075406.1 | - | 9 - 10 | 1046 | 92 | |
CR760768.2 | - | 9 - 10 | 1048 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |