Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006716.1 | NP_001006717.1 | 4 - 5 | 701 | 69 | |
XM_012966388.2 | XP_012821842.1 | 4 - 5 | 723 | 69 | ||
GenBank mRNA | BC075445.1 | - | 4 - 5 | 701 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |