Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006719.1 | NP_001006720.1 | 3 - 4 | 455 | 75 | |
XM_012959567.1 | XP_012815021.1 | 2 - 3 | 287 | 75 | ||
GenBank mRNA | BC075454.1 | - | 3 - 4 | 455 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |