Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006763.1 | NP_001006764.1 | 3 - 4 | 164 | 77 | |
XM_012968229.2 | XP_012823683.1 | 3 - 4 | 231 | 77 | ||
XM_012968231.2 | XP_012823685.1 | 3 - 4 | 217 | 77 | ||
XM_012968232.2 | XP_012823686.1 | 3 - 4 | 236 | 77 | ||
GenBank mRNA | BC075555.1 | - | 3 - 4 | 164 | 77 | |
CR761647.2 | - | 3 - 4 | 170 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |