Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006768.1 | NP_001006769.1 | 10 - 11 | 924 | 86 | |
GenBank mRNA | BC075563.1 | - | 10 - 11 | 924 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |