Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctdsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006792.1 | NP_001006793.1 | 4 - 5 | 747 | 76 | |
XM_012956751.2 | XP_012812205.1 | 5 - 6 | 1209 | 76 | ||
XM_018091166.1 | XP_017946655.1 | 3 - 4 | 1285 | 76 | ||
GenBank mRNA | BC076658.1 | - | 4 - 5 | 747 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |