Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gstt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006810.1 | NP_001006811.1 | 2 - 3 | 217 | 87 | |
GenBank mRNA | BC076690.1 | - | 2 - 3 | 217 | 87 | |
CR760091.2 | - | 2 - 3 | 224 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |