Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006817.1 | NP_001006818.1 | 5 - 6 | 815 | 62 | |
GenBank mRNA | BC076712.1 | - | 5 - 6 | 815 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |