Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smarcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006818.1 | NP_001006819.1 | 8 - 9 | 1095 | 75 | |
XM_012959983.2 | XP_012815437.1 | 8 - 9 | 1134 | 75 | ||
GenBank mRNA | BC076714.1 | - | 8 - 9 | 1095 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |