Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006825.1 | NP_001006826.1 | 3 - 4 | 615 | 70 | |
GenBank mRNA | BC076890.1 | - | 3 - 4 | 615 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |