Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006835.1 | NP_001006836.1 | 4 - 5 | 477 | 84 | |
XM_012960867.2 | XP_012816321.1 | 4 - 5 | 530 | 84 | ||
GenBank mRNA | BC076911.1 | - | 4 - 5 | 477 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |