Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006862.1 | NP_001006863.1 | 1 - 2 | 216 | 66 | |
GenBank mRNA | BC076971.1 | - | 1 - 2 | 216 | 66 | |
CR855423.2 | - | 1 - 2 | 125 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |