Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006862.1 | NP_001006863.1 | 3 - 4 | 515 | 60 | |
GenBank mRNA | BC076971.1 | - | 3 - 4 | 515 | 60 | |
CR855423.2 | - | 3 - 4 | 424 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |