Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006864.1 | NP_001006865.1 | 1 - 2 | 209 | 94 | |
XM_012956765.2 | XP_012812219.1 | 1 - 2 | 558 | 94 | ||
XM_012956766.2 | XP_012812220.1 | 1 - 2 | 558 | 94 | ||
XM_012956767.2 | XP_012812221.1 | 1 - 2 | 559 | 94 | ||
GenBank mRNA | BC076974.1 | - | 1 - 2 | 209 | 94 | |
CT030354.1 | - | 1 - 2 | 140 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |