Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006869.1 | NP_001006870.1 | 2 - 3 | 554 | 65 | |
GenBank mRNA | BC076983.1 | - | 2 - 3 | 554 | 65 | |
KJ617083.1 | - | 2 - 3 | 336 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |