Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006900.1 | NP_001006901.1 | 5 - 6 | 604 | 86 | |
XM_012960212.2 | XP_012815666.1 | 5 - 6 | 814 | 86 | ||
XM_012960215.2 | XP_012815669.1 | 4 - 5 | 769 | 86 | ||
XM_018092426.1 | XP_017947915.1 | 5 - 6 | 776 | 86 | ||
XM_018092431.1 | XP_017947920.1 | 5 - 6 | 773 | 86 | ||
XM_018092433.1 | XP_017947922.1 | 4 - 5 | 715 | 86 | ||
GenBank mRNA | BC074695.1 | - | 5 - 6 | 604 | 86 | |
CR926179.2 | - | 5 - 6 | 725 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |