Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006900.1 | NP_001006901.1 | 7 - 8 | 955 | 88 | |
XM_012960212.2 | XP_012815666.1 | 7 - 8 | 1165 | 88 | ||
XM_012960215.2 | XP_012815669.1 | 6 - 7 | 1120 | 88 | ||
XM_018092426.1 | XP_017947915.1 | 7 - 8 | 1127 | 88 | ||
XM_018092431.1 | XP_017947920.1 | 7 - 8 | 1124 | 88 | ||
XM_018092433.1 | XP_017947922.1 | 6 - 7 | 1066 | 88 | ||
GenBank mRNA | BC074695.1 | - | 7 - 8 | 955 | 88 | |
CR926179.2 | - | 7 - 8 | 1076 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |