Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006913.1 | NP_001006914.1 | 9 - 10 | 878 | 87 | |
XM_018093843.1 | XP_017949332.1 | 10 - 11 | 821 | 87 | ||
XM_018093844.1 | XP_017949333.1 | 10 - 11 | 814 | 87 | ||
GenBank mRNA | BC075395.1 | - | 9 - 10 | 878 | 87 | |
BC087758.1 | - | 9 - 10 | 803 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |