Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006915.1 | NP_001006916.1 | 3 - 4 | 669 | 63 | |
GenBank mRNA | BC075431.1 | - | 3 - 4 | 669 | 63 | |
CR848225.2 | - | 3 - 4 | 670 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |