Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007509.1 | NP_001007510.1 | 11 - 12 | 1377 | 76 | |
XM_012956782.2 | XP_012812236.1 | 12 - 13 | 1402 | 76 | ||
GenBank mRNA | BC079946.1 | - | 11 - 12 | 1377 | 76 | |
CT030112.2 | - | 11 - 12 | 1349 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |