Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007513.1 | NP_001007514.1 | 7 - 8 | 1082 | 66 | |
XM_012958864.1 | XP_012814318.1 | 7 - 8 | 1144 | 66 | ||
XM_012958865.1 | XP_012814319.1 | 7 - 8 | 1141 | 66 | ||
XM_012958867.2 | XP_012814321.1 | 7 - 8 | 1124 | 66 | ||
XM_012958868.2 | XP_012814322.1 | 7 - 8 | 1114 | 66 | ||
XM_012958869.1 | XP_012814323.1 | 7 - 8 | 941 | 66 | ||
XM_012958870.2 | XP_012814324.1 | 7 - 8 | 958 | 66 | ||
XM_012958871.2 | XP_012814325.1 | 7 - 8 | 943 | 66 | ||
GenBank mRNA | BC079954.1 | - | 7 - 8 | 1082 | 66 | |
CT030587.1 | - | 7 - 8 | 923 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |