Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: surf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007864.1 | NP_001007865.1 | 2 - 3 | 189 | 69 | |
XM_012968261.2 | XP_012823715.1 | 2 - 3 | 203 | 69 | ||
GenBank mRNA | BC079967.1 | - | 2 - 3 | 189 | 69 | |
CR926263.1 | - | 2 - 3 | 207 | 69 | ||
CR942359.2 | - | 1 - 2 | 187 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |