Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007872.1 | NP_001007873.1 | 3 - 4 | 294 | 103 | |
XM_012960249.2 | XP_012815703.1 | 3 - 4 | 372 | 103 | ||
XM_012960250.2 | XP_012815704.1 | 3 - 4 | 521 | 103 | ||
XM_012960257.2 | XP_012815711.1 | 3 - 4 | 457 | 103 | ||
GenBank mRNA | BC080139.1 | - | 3 - 4 | 294 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |