Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tekt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007883.2 | NP_001007884.1 | 5 - 6 | 690 | 74 | |
GenBank mRNA | BC080153.1 | - | 5 - 6 | 692 | 74 | |
CR760301.2 | - | 5 - 6 | 725 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |