Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007932.1 | NP_001007933.1 | 2 - 3 | 429 | 78 | |
XM_012970321.2 | XP_012825775.1 | 1 - 2 | 172 | 78 | ||
GenBank mRNA | BC080440.1 | - | 2 - 3 | 429 | 78 | |
CR761660.2 | - | 1 - 2 | 113 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |