Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ints14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007938.1 | NP_001007939.1 | 10 - 11 | 1385 | 99 | |
XM_012958881.2 | XP_012814335.1 | 10 - 11 | 1819 | 99 | ||
GenBank mRNA | BC080444.1 | - | 10 - 11 | 1385 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |