Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uevld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007957.1 | NP_001007958.1 | 1 - 2 | 203 | 75 | |
GenBank mRNA | BC080471.1 | - | 1 - 2 | 203 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |