Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007968.2 | NP_001007969.2 | 4 - 5 | 660 | 65 | |
GenBank mRNA | BC080480.1 | - | 4 - 5 | 661 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |