Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007984.1 | NP_001007985.1 | 2 - 3 | 286 | 109 | |
XM_018091189.1 | XP_017946678.1 | 3 - 4 | 730 | 109 | ||
XM_018091190.1 | XP_017946679.1 | 4 - 5 | 800 | 109 | ||
GenBank mRNA | BC080494.1 | - | 2 - 3 | 286 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |