Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008012.1 | NP_001008013.1 | 3 - 4 | 786 | 92 | |
XM_012971669.2 | XP_012827123.1 | 3 - 4 | 852 | 92 | ||
GenBank mRNA | BC080891.1 | - | 3 - 4 | 786 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |