Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008036.1 | NP_001008037.1 | 2 - 3 | 333 | 59 | |
GenBank mRNA | AY714338.1 | - | 2 - 3 | 333 | 59 | |
BC080919.1 | - | 2 - 3 | 333 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |