Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008070.1 | NP_001008071.1 | 3 - 4 | 251 | 89 | |
XM_012960935.2 | XP_012816389.1 | 2 - 3 | 683 | 89 | ||
XM_018093019.1 | XP_017948508.1 | 3 - 4 | 703 | 89 | ||
GenBank mRNA | BC080964.1 | - | 3 - 4 | 251 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |