Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008070.1 | NP_001008071.1 | 5 - 6 | 409 | 82 | |
XM_012960935.2 | XP_012816389.1 | 4 - 5 | 841 | 82 | ||
XM_018093019.1 | XP_017948508.1 | 5 - 6 | 861 | 82 | ||
GenBank mRNA | BC080964.1 | - | 5 - 6 | 409 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |