Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008072.1 | NP_001008073.1 | 2 - 3 | 298 | 88 | |
GenBank mRNA | BC080967.1 | - | 2 - 3 | 298 | 88 | |
CT030458.1 | - | 242 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |