Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctrc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008076.1 | NP_001008077.1 | 3 - 4 | 242 | 58 | |
GenBank mRNA | BC080976.1 | - | 3 - 4 | 242 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |