Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008092.1 | NP_001008093.1 | 3 - 4 | 668 | 63 | |
GenBank mRNA | BC081291.1 | - | 3 - 4 | 668 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |