Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008107.1 | NP_001008108.1 | 1 - 2 | 244 | 68 | |
GenBank mRNA | BC081310.1 | - | 1 - 2 | 244 | 68 | |
CR762151.2 | - | 1 - 2 | 260 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |