Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008107.1 | NP_001008108.1 | 2 - 3 | 373 | 107 | |
GenBank mRNA | BC081310.1 | - | 2 - 3 | 373 | 107 | |
CR762151.2 | - | 2 - 3 | 389 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |