Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nup35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008123.1 | NP_001008124.1 | 3 - 4 | 388 | 70 | |
XM_012970344.2 | XP_012825798.1 | 3 - 4 | 484 | 70 | ||
XM_012970345.2 | XP_012825799.1 | 3 - 4 | 474 | 70 | ||
XM_012970346.1 | XP_012825800.1 | 3 - 4 | 419 | 70 | ||
GenBank mRNA | BC081329.1 | - | 3 - 4 | 388 | 70 | |
CR761020.2 | - | 3 - 4 | 321 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |