Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008153.1 | NP_001008154.1 | 2 - 3 | 460 | 80 | |
GenBank mRNA | BC081367.1 | - | 2 - 3 | 460 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |