Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kpna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008154.2 | NP_001008155.1 | 5 - 6 | 635 | 69 | |
XM_012966478.2 | XP_012821932.1 | 4 - 5 | 697 | 69 | ||
XM_012966479.2 | XP_012821933.1 | 5 - 6 | 728 | 69 | ||
GenBank mRNA | BC081368.1 | - | 5 - 6 | 637 | 69 | |
CR762026.2 | - | 4 - 5 | 608 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |