Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ywhae | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008155.1 | NP_001008156.1 | 4 - 5 | 754 | 104 | |
XM_012956823.1 | XP_012812277.1 | 4 - 5 | 821 | 104 | ||
GenBank mRNA | BC081369.1 | - | 4 - 5 | 754 | 104 | |
CR848241.2 | - | 4 - 5 | 614 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |