Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008159.1 | NP_001008160.1 | 5 - 6 | 506 | 68 | |
GenBank mRNA | BC082340.1 | - | 5 - 6 | 506 | 68 | |
CR760752.1 | - | 5 - 6 | 499 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |