Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: upp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008173.1 | NP_001008174.1 | 1 - 2 | 151 | 72 | |
GenBank mRNA | BC082493.1 | - | 1 - 2 | 151 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |