Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008176.1 | NP_001008177.1 | 2 - 3 | 483 | 83 | |
XM_012966482.2 | XP_012821936.1 | 3 - 4 | 499 | 83 | ||
XM_012966483.2 | XP_012821937.1 | 3 - 4 | 495 | 83 | ||
XM_012966484.1 | XP_012821938.1 | 3 - 4 | 384 | 83 | ||
XM_012966485.2 | XP_012821939.1 | 2 - 3 | 599 | 83 | ||
GenBank mRNA | BC082496.1 | - | 2 - 3 | 483 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |