Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bloc1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008200.1 | NP_001008201.1 | 4 - 5 | 400 | 98 | |
XM_012966489.2 | XP_012821943.1 | 4 - 5 | 456 | 98 | ||
XM_012966490.2 | XP_012821944.1 | 4 - 5 | 502 | 98 | ||
GenBank mRNA | BC080469.1 | - | 4 - 5 | 400 | 98 | |
CR760883.2 | - | 4 - 5 | 401 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |