Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008204.1 | NP_001008205.1 | 7 - 8 | 749 | 103 | |
XM_012968277.2 | XP_012823731.1 | 7 - 8 | 725 | 103 | ||
XM_012968278.2 | XP_012823732.1 | 7 - 8 | 806 | 103 | ||
GenBank mRNA | BC080963.1 | - | 7 - 8 | 749 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |