Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008206.1 | NP_001008207.1 | 505 | 76 | ||
GenBank mRNA | BC081292.1 | - | 505 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis |