Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008439.1 | NP_001008439.1 | 10 - 11 | 1007 | 85 | |
GenBank mRNA | BC059755.1 | - | 10 - 11 | 981 | 85 | |
BC080337.1 | - | 10 - 11 | 1007 | 85 | ||
CR761691.2 | - | 10 - 11 | 964 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |