Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008439.1 | NP_001008439.1 | 2 - 3 | 284 | 101 | |
GenBank mRNA | BC059755.1 | - | 2 - 3 | 258 | 101 | |
BC080337.1 | - | 2 - 3 | 284 | 101 | ||
CR761691.2 | - | 2 - 3 | 241 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |